Methyl-4-(4-methoxyphenyl)-2-oxo-3-butenoate ≥95%

Supplier: Matrix Scientific

048252-500MG 048252-1G
101883-480EA 172.31 USD
101883-480 101883-482
Methyl-4-(4-methoxyphenyl)-2-oxo-3-butenoate ≥95%
Methyl-4-(4-methoxyphenyl)-2-oxo-3-butenoate

Hazard: Irritant

Formula: C₁₂H₁₂O₄
MW: 220.23 g/mol
MDL Number: MFCD09864843
CAS Number: 105213-31-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR