4-(Phenylsulfanyl)aniline ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(Phenylthio)aniline

048251-500MG
101883-478EA 92.31 USD
101883-478
4-(Phenylsulfanyl)aniline ≥95%
4-(Phenylsulfanyl)aniline

Hazard: Irritant

Formula: C₁₂H₁₁NS
MW: 201.29 g/mol
MDL Number: MFCD00094027
CAS Number: 1135-14-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR