Methyl-2-(4-chlorophenyl)-2-methylpropanoate ≥95%
Supplier: Matrix Scientific
048249-500MG
101883-472EA
181.54
USD
101883-472
Methyl-2-(4-chlorophenyl)-2-methylpropanoate ≥95%
Methyl-2-(4-chlorophenyl)-2-methylpropanoate
Hazard: Irritant
Formula:
C₁₁H₁₃ClO₂ MW: 212.68 g/mol |
MDL Number:
MFCD09865030 CAS Number: 57225-86-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...