2-(3,4-Dihydro-2H-pyrrol-5-yl)-1H-pyrrole ≥95%
Supplier: Matrix Scientific
Synonyms:
4',5'-Dihydro-1H,3'H-2,2'-bipyrrole
048243-1G
048243-500MG
101883-448EA
375.38
USD
101883-448
101883-450
2-(3,4-Dihydro-2H-pyrrol-5-yl)-1H-pyrrole ≥95%
2-(3,4-Dihydro-2H-pyrrol-5-yl)-1H-pyrrole
Hazard: Irritant
Formula:
C₈H₁₀N₂ MW: 134.18 g/mol |
MDL Number:
MFCD00030437 CAS Number: 10087-65-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...