2-Methyl-2-(2-thienyl)butanoic acid ≥95%
Supplier: Matrix Scientific
048239-500MG
101883-430EA
556.92
USD
101883-430
2-Methyl-2-(2-thienyl)butanoic acid ≥95%
2-Methyl-2-(2-thienyl)butanoic acid
Hazard: Irritant
Formula:
C₉H₁₂O₂S MW: 184.26 g/mol |
MDL Number:
MFCD09865028 CAS Number: 1017783-11-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...