tert-Butyl-4-(3-cyano-6-cyclopropyl-2-pyridinyl)tetrahydro-1(2H)-pyrazinecarboxylate ≥95%

Supplier: Matrix Scientific

048221-500MG 048221-1G
101883-348EA 556.92 USD
101883-348 101883-350
tert-Butyl-4-(3-cyano-6-cyclopropyl-2-pyridinyl)tetrahydro-1(2H)-pyrazinecarboxylate ≥95%
tert-Butyl-4-(3-cyano-6-cyclopropyl-2-pyridinyl)tetrahydro-1(2H)-pyrazinecarboxylate

Hazard: Irritant

Formula: C₁₈H₂₄N₄O₂
MW: 328.42 g/mol
MDL Number: MFCD11841055
CAS Number: 1135283-84-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR