4-(5-Isopropyl-1,2,4-oxadiazol-3-yl)-2-piperazin-1-ylpyrimidine dihydrochloride ≥95%

Supplier: Matrix Scientific

048192-500MG
101883-248EA 1109.23 USD
101883-248
4-(5-Isopropyl-1,2,4-oxadiazol-3-yl)-2-piperazin-1-ylpyrimidine dihydrochloride ≥95%
4-(5-Isopropyl-1,2,4-oxadiazol-3-yl)-2-piperazin-1-ylpyrimidine dihydrochloride

Hazard: Irritant

Formula: C₁₃H₂₀Cl₂N₆O
MW: 347.25 g/mol
MDL Number: MFCD12025943
CAS Number: 1177093-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR