(2-Morpholino-5-nitrophenyl)methanol ≥97%
Supplier: Matrix Scientific
048181-5G
048181-500MG
048181-1G
101883-202EA
350.77
USD
101883-202
101883-204
101883-200
(2-Morpholino-5-nitrophenyl)methanol ≥97%
(2-Morpholino-5-nitrophenyl)methanol
Hazard: Irritant
Formula:
C₁₁H₁₄N₂O₄ |
MDL Number:
MFCD01312994 CAS Number: 300665-25-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...