2-Benzyloctahydro-1H-isoindol-4-amine ≥95%

Supplier: Matrix Scientific

048162-500MG
101883-104EA 1483.08 USD
101883-104
2-Benzyloctahydro-1H-isoindol-4-amine ≥95%
2-Benzyloctahydro-1H-isoindol-4-amine

Hazard: Irritant

Formula: C₁₅H₂₂N₂
MDL Number: MFCD10758104
CAS Number: 217464-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR