4-(2,4-Dinitrophenyl)butanoic acid ≥95%

Supplier: Matrix Scientific

048120-500MG 048120-1G 048120-5G
101882-896EA 579.6 USD
101882-896 101882-894 101882-892
4-(2,4-Dinitrophenyl)butanoic acid ≥95%
4-(2,4-Dinitrophenyl)butanoic acid

Hazard: Irritant

Formula: C₁₀H₁₀N₂O₆
MDL Number: MFCD05663835
CAS Number: 52120-49-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR