4-(tert-Butyl)-2-methyl-(2R,4R,5S)-5-(4-methoxyphenyl)tetrahydro-1H-pyrrole-2,4-dicarboxylate ≥95%

Supplier: Matrix Scientific

048086-500MG
101882-742EA 747.69 USD
101882-742
4-(tert-Butyl)-2-methyl-(2R,4R,5S)-5-(4-methoxyphenyl)tetrahydro-1H-pyrrole-2,4-dicarboxylate ≥95%
4-(tert-Butyl)-2-methyl-(2R,4R,5S)-5-(4-methoxyphenyl)tetrahydro-1H-pyrrole-2,4-dicarboxylate

Hazard: Irritant

Formula: C₁₈H₂₅NO₅
MDL Number: MFCD11841023
CAS Number: 1219423-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR