2-Benzyloctahydrocyclopenta[c]pyrrol-4-ylamine ≥95%

Supplier: Matrix Scientific

048039-500MG
101882-504EA 1483.08 USD
101882-504
2-Benzyloctahydrocyclopenta[c]pyrrol-4-ylamine ≥95%
2-Benzyloctahydrocyclopenta[c]pyrrol-4-ylamine

Hazard: Irritant

Formula: C₁₄H₂₀N₂
MDL Number: MFCD06658344
CAS Number: 186201-60-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR