3-(4-Bromophenyl)-5,6-dihydroimidazo[2,1-b][1,3]thiazole-2-carbaldehyde ≥95%

Supplier: Matrix Scientific

048038-500MG
101882-502EA 556.92 USD
101882-502
3-(4-Bromophenyl)-5,6-dihydroimidazo[2,1-b][1,3]thiazole-2-carbaldehyde ≥95%
3-(4-Bromophenyl)-5,6-dihydroimidazo[2,1-b][1,3]thiazole-2-carbaldehyde

Hazard: Irritant

Formula: C₁₂H₉BrN₂OS
MDL Number: MFCD10758103
CAS Number: 1033463-44-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR