1-Allyltetrahydro-4(1H)-pyridinone ≥95%

Supplier: Matrix Scientific

048027-1G 048027-5G 048027-500MG
101882-454EA 806.15 USD
101882-454 101882-452 101882-450
1-Allyltetrahydro-4(1H)-pyridinone ≥95%
1-Allyltetrahydro-4(1H)-pyridinone

Hazard: Irritant

Formula: C₈H₁₃NO
MDL Number: MFCD09152746
CAS Number: 20785-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR