4-Hydrazinoquinazoline ≥95%

Supplier: Matrix Scientific

047962-500MG 047962-1G
101882-182EA 375.38 USD
101882-182 101882-184
4-Hydrazinoquinazoline ≥95%
4-Hydrazinoquinazoline

Hazard: Irritant

Formula: C₈H₈N₄
MDL Number: MFCD01219546
CAS Number: 36075-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR