5-Bromo-2-chloro-3-(chloromethyl)pyridine ≥95%

Supplier: Matrix Scientific

047943-500MG
101882-096EA 829.15 USD
101882-096
5-Bromo-2-chloro-3-(chloromethyl)pyridine ≥95%
5-Bromo-2-chloro-3-(chloromethyl)pyridine

Hazard: Irritant

Formula: C₆H₄BrCl₂N
MDL Number: MFCD09865020
CAS Number: 1017782-64-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR