2-Benzylhexahydrocyclopenta[c]pyrrol-4(1H)-one ≥95%
Supplier: Matrix Scientific
047926-1G
047926-500MG
101882-032EA
927.69
USD
101882-032
101882-030
2-Benzylhexahydrocyclopenta[c]pyrrol-4(1H)-one ≥95%
2-Benzylhexahydrocyclopenta[c]pyrrol-4(1H)-one
Hazard: Irritant
Formula:
C₁₄H₁₇NO MW: 215.3 g/mol |
MDL Number:
MFCD06658339 CAS Number: 185692-51-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...