Dobutamine hydrochloride ≥98%
Supplier: Matrix Scientific
Synonyms:
3,4-Dihydroxy-N-[3-(4-hydroxyphenyl)-1-methylpropyl]-beta-phenethylamine Hydrochloride
047888-1G
047888-500MG
047888-5G
101881-864EA
747.69
USD
101881-864
101881-866
101881-868
Dobutamine hydrochloride ≥98%
Dobutamine hydrochloride
Purity:
CAS: 49745-95-1
MDL Number: MFCD00153795
Molecular Formula: C18H24ClNO3
Molecular Weight: 337,85
Hazard: Irritant
Formula:
C₁₈H₂₃NO₃ MW: 337.84 g/mol Melting Pt: 184…186 °C |
MDL Number:
MFCD00153795 CAS Number: 49745-95-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...