4-[5-(4-Methylphenyl)-3-(trifluoromethyl)pyrazol-1-yl]benzenesulfonamide ≥98%

Supplier: Matrix Scientific

Synonyms: Celecoxib

047884-5G
101881-848EA 89.7 USD
101881-848
4-[5-(4-Methylphenyl)-3-(trifluoromethyl)pyrazol-1-yl]benzenesulfonamide ≥98%
4-[5-(4-Methylphenyl)-3-(trifluoromethyl)pyrazol-1-yl]benzenesulfonamide

Hazard: Irritant

Formula: C₁₇H₁₄F₃N₃O₂S
MW: 381.38 g/mol
MDL Number: MFCD00941298
CAS Number: 169590-42-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR