3-Fluoro-4-methylthiophenol ≥95%
Supplier: Matrix Scientific
047795-1G
047795-5G
047795-500MG
101881-472EA
375.38
USD
101881-472
101881-474
101881-476
3-Fluoro-4-methylthiophenol ≥95%
3-Fluoro-4-methylthiophenol
Hazard: Irritant
Formula:
C₇H₇FS |
MDL Number:
MFCD06201750 CAS Number: 64359-35-9 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...