tert-Butyl-2-(2-ethoxy-2-oxoethyl)tetrahydro-1(2H)-pyridazinecarboxylate ≥95%
Supplier: Matrix Scientific
046374-500MG
101875-222EA
1109.23
USD
101875-222
tert-Butyl-2-(2-ethoxy-2-oxoethyl)tetrahydro-1(2H)-pyridazinecarboxylate ≥95%
tert-Butyl-2-(2-ethoxy-2-oxoethyl)tetrahydro-1(2H)-pyridazinecarboxylate
Hazard: Irritant
Formula:
C₁₃H₂₄N₂O₄ MW: 272.35 g/mol |
MDL Number:
MFCD09152723 CAS Number: 952183-06-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...