Diethyl-2-[(4-methylphenyl)methylene]malonate ≥95%

Supplier: Matrix Scientific

046342-500MG
101875-076EA 280.6 USD
101875-076
Diethyl-2-[(4-methylphenyl)methylene]malonate ≥95%
Diethyl-2-[(4-methylphenyl)methylene]malonate

Hazard: Irritant

Formula: C₁₅H₁₈O₄
MDL Number: MFCD00524854
CAS Number: 14111-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR