N,N-Diethyl-3-phenylacrylamide ≥95%

Supplier: Matrix Scientific

046341-500MG
101875-074EA 208.15 USD
101875-074
N,N-Diethyl-3-phenylacrylamide ≥95%
N,N-Diethyl-3-phenylacrylamide

Hazard: Irritant

Formula: C₁₃H₁₇NO
Melting Pt: 69…70 °C
MDL Number: MFCD00048528
CAS Number: 3680-04-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR