3,5-Difluoro-4-pyridinamine ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Amino-3,5-difluoropyridine3,5-Difluoropyridin-4-amine

046330-1G
101875-052EA 69.23 USD
101875-052
3,5-Difluoro-4-pyridinamine ≥95%
3,5-Difluoro-4-pyridinamine

Hazard: Irritant

Formula: C₅H₄F₂N₂
MW: 130.1 g/mol
MDL Number: MFCD01862101
CAS Number: 159783-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR