N-(3-Chloro-4-methylphenyl)methanesulfonamide ≥95%

Supplier: Matrix Scientific

046327-500MG
101875-044EA 208.15 USD
101875-044
N-(3-Chloro-4-methylphenyl)methanesulfonamide ≥95%
N-(3-Chloro-4-methylphenyl)methanesulfonamide

Hazard: Irritant

Formula: C₈H₁₀ClNO₂S
MDL Number: MFCD01212924
CAS Number: 71270-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR