1-(3,4-Dimethoxyphenyl)-8,9-dimethoxy-3-phenylpyrrolo[2,1-a]isoquinoline ≥95%

Supplier: Matrix Scientific

046325-500MG
101875-040EA 1109.23 USD
101875-040
1-(3,4-Dimethoxyphenyl)-8,9-dimethoxy-3-phenylpyrrolo[2,1-a]isoquinoline ≥95%
1-(3,4-Dimethoxyphenyl)-8,9-dimethoxy-3-phenylpyrrolo[2,1-a]isoquinoline

Hazard: Irritant

Formula: C₂₈H₂₅NO₄
MDL Number: MFCD08056693
CAS Number: 886362-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR