4-Hydroxy-8-methoxy-2-(trifluoromethyl)quinoline ≥95%

Supplier: Matrix Scientific

046312-500MG
101875-014EA 416.3 USD
101875-014
4-Hydroxy-8-methoxy-2-(trifluoromethyl)quinoline ≥95%
4-Hydroxy-8-methoxy-2-(trifluoromethyl)quinoline

Hazard: Irritant

Formula: C₁₁H₈F₃NO₂
MDL Number: MFCD01862071
CAS Number: 41192-84-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR