2,5-Dimethylphenyl acetate ≥95%

Supplier: Matrix Scientific

046306-500MG
101875-002EA 208.15 USD
101875-002
2,5-Dimethylphenyl acetate ≥95%
2,5-Dimethylphenyl acetate

Hazard: Irritant

Formula: C₁₀H₁₂O₂
MDL Number: MFCD03357477
CAS Number: 877-48-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR