3,6-Bis[(chlorocarbonyl)oxy]hexahydrofuro[3,2-b]furan ≥95%

Supplier: Matrix Scientific

046303-500MG
101874-996EA 278.46 USD
101874-996
3,6-Bis[(chlorocarbonyl)oxy]hexahydrofuro[3,2-b]furan ≥95%
3,6-Bis[(chlorocarbonyl)oxy]hexahydrofuro[3,2-b]furan

Hazard: Irritant

Formula: C₈H₈Cl₂O₆
MW: 271.06 g/mol
MDL Number: MFCD08689671
CAS Number: 163877-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR