4-(Hydroxymethyl)-1-methylpyridinium iodide ≥95%

Supplier: Matrix Scientific

046302-500MG
101874-994EA 384.62 USD
101874-994
4-(Hydroxymethyl)-1-methylpyridinium iodide ≥95%
4-(Hydroxymethyl)-1-methylpyridinium iodide

Hazard: Irritant

Formula: C₇H₁₀INO
MDL Number: MFCD00956041
CAS Number: 6457-57-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR