5-Allyl-2-amino-6-chloro-4-pyrimidinol ≥95%

Supplier: Matrix Scientific

046299-500MG
101874-990EA 556.92 USD
101874-990
5-Allyl-2-amino-6-chloro-4-pyrimidinol ≥95%
5-Allyl-2-amino-6-chloro-4-pyrimidinol

Hazard: Irritant

Formula: C₇H₈ClN₃O
MDL Number: MFCD08056684
CAS Number: 886362-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR