1-(Methoxymethoxy)benzene ≥95%

Supplier: Matrix Scientific

046297-500MG
101874-984EA 653.85 USD
101874-984
1-(Methoxymethoxy)benzene ≥95%
1-(Methoxymethoxy)benzene

Hazard: Irritant

Formula: C₈H₁₀O₂
MDL Number: MFCD00457267
CAS Number: 824-91-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR