2,2-Dimethoxy-N-(phenylmethylene)-1-ethanamine ≥95%

Supplier: Matrix Scientific

046289-500MG
101874-964EA 278.46 USD
101874-964
2,2-Dimethoxy-N-(phenylmethylene)-1-ethanamine ≥95%
2,2-Dimethoxy-N-(phenylmethylene)-1-ethanamine

Hazard: Irritant

Formula: C₁₁H₁₅NO₂
MDL Number: MFCD08056680
CAS Number: 54879-74-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR