5-Chloro-6-cyclopropyl-1-methyl-1H-pyrazolo[3,4-b]pyridin-3-amine ≥95%
Supplier: Matrix Scientific
046286-500MG
046286-1G
101874-956EA
869.4
USD
101874-956
101874-954
5-Chloro-6-cyclopropyl-1-methyl-1H-pyrazolo[3,4-b]pyridin-3-amine ≥95%
5-Chloro-6-cyclopropyl-1-methyl-1H-pyrazolo[3,4-b]pyridin-3-amine
Hazard: Irritant
Formula:
C₁₀H₁₁ClN₄ |
MDL Number:
MFCD12025852 CAS Number: 1135283-21-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...