1-Benzyl-5-nitro-1H-1,3-benzimidazole ≥95%

Supplier: Matrix Scientific

046261-1G
101874-864EA 349.6 USD
101874-864
1-Benzyl-5-nitro-1H-1,3-benzimidazole ≥95%
1-Benzyl-5-nitro-1H-1,3-benzimidazole

Hazard: Irritant

Formula: C₁₄H₁₁N₃O₂
MDL Number: MFCD00837324
CAS Number: 15207-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR