5-Nitro-2-piperidinoaniline ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Nitro-2-(piperidin-1-yl)aniline

046235-500MG 046235-1G
101874-774EA 262.2 USD
101874-774 101874-772
5-Nitro-2-piperidinoaniline ≥95%
5-Nitro-2-piperidinoaniline

Hazard: Irritant

Formula: C₁₁H₁₅N₃O₂
MW: 221.26 g/mol
MDL Number: MFCD00852821
CAS Number: 5367-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR