4-(tert-Butyl)-2-methyl-(2R,5S)-2-isobutyl-5-(1,3-thiazol-2-yl)tetrahydro-1H-pyrrole-2,4-dicarboxylate ≥95%

Supplier: Matrix Scientific

046226-500MG
101874-736EA 1109.23 USD
101874-736
4-(tert-Butyl)-2-methyl-(2R,5S)-2-isobutyl-5-(1,3-thiazol-2-yl)tetrahydro-1H-pyrrole-2,4-dicarboxylate ≥95%
4-(tert-Butyl)-2-methyl-(2R,5S)-2-isobutyl-5-(1,3-thiazol-2-yl)tetrahydro-1H-pyrrole-2,4-dicarboxylate

Hazard: Irritant

Formula: C₁₈H₂₈N₂O₄S
MDL Number: MFCD12025840
CAS Number: 1217691-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR