4-(5-Cyclopropyl-1,2,4-oxadiazol-3-yl)-2-piperazin-1-ylpyrimidine dihydrochloride ≥95%

Supplier: Matrix Scientific

046219-500MG
101874-718EA 829.15 USD
101874-718
4-(5-Cyclopropyl-1,2,4-oxadiazol-3-yl)-2-piperazin-1-ylpyrimidine dihydrochloride ≥95%
4-(5-Cyclopropyl-1,2,4-oxadiazol-3-yl)-2-piperazin-1-ylpyrimidine dihydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈Cl₂N₆O
MDL Number: MFCD12025836
CAS Number: 1177093-08-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR