Ethyl-1-(2-chloro-2,2-difluoroacetyl)-4-piperidinecarboxylate ≥95%
Supplier: Matrix Scientific
045910-500MG
045910-5G
045910-1G
101873-212EA
181.54
USD
101873-212
101873-214
101873-216
Ethyl-1-(2-chloro-2,2-difluoroacetyl)-4-piperidinecarboxylate ≥95%
Ethyl-1-(2-chloro-2,2-difluoroacetyl)-4-piperidinecarboxylate
Hazard: Irritant
Formula:
C₁₀H₁₄ClF₂NO₃ |
MDL Number:
MFCD02082447 CAS Number: 454473-81-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...