Ethyl-2-cyano-3-[3-(trifluoromethyl)-phenyl]acrylate ≥97%

Supplier: Matrix Scientific

045901-1G 045901-500MG
101873-180EA 232.31 USD
101873-180 101873-178
Ethyl-2-cyano-3-[3-(trifluoromethyl)-phenyl]acrylate ≥97%
Ethyl-2-cyano-3-[3-(trifluoromethyl)-phenyl]acrylate

Hazard: Irritant

Formula: C₁₃H₁₀F₃NO₂
MW: 269.23 g/mol
MDL Number: MFCD03283527
CAS Number: 149550-20-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR