Ethyl-2-cyano-3-(4-pyridinyl)acrylate ≥95%
Supplier: Matrix Scientific
045900-5G
045900-1G
045900-500MG
101873-172EA
132.31
USD
101873-172
101873-174
101873-176
Ethyl-2-cyano-3-(4-pyridinyl)acrylate ≥95%
Ethyl-2-cyano-3-(4-pyridinyl)acrylate
Hazard: Irritant
Formula:
C₁₁H₁₀N₂O₂ MW: 202.21 g/mol |
MDL Number:
MFCD00175069 CAS Number: 123293-73-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...