6-(4-Propoxyphenoxy)nicotinonitrile ≥95%

Supplier: Matrix Scientific

044373-500MG 044373-1G 044373-5G
101865-512EA 278.46 USD
101865-512 101865-516 101865-514
6-(4-Propoxyphenoxy)nicotinonitrile ≥95%
6-(4-Propoxyphenoxy)nicotinonitrile

Hazard: Irritant

Formula: C₁₅H₁₄N₂O₂
MDL Number: MFCD10693376
CAS Number: 1135282-78-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR