5-(1-Methylhydrazino)-1,2,3-thiadiazole ≥95%

Supplier: Matrix Scientific

044123-500MG
101864-260EA 829.15 USD
101864-260
5-(1-Methylhydrazino)-1,2,3-thiadiazole ≥95%
5-(1-Methylhydrazino)-1,2,3-thiadiazole

Hazard: Irritant

Formula: C₃H₆N₄S
MDL Number: MFCD12025829
CAS Number: 1147979-33-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR