2-(2-Chloro-3-pyridinyl)-1H-1,3-benzimidazole ≥95%

Supplier: Matrix Scientific

044114-500MG
101864-218EA 556.92 USD
101864-218
2-(2-Chloro-3-pyridinyl)-1H-1,3-benzimidazole ≥95%
2-(2-Chloro-3-pyridinyl)-1H-1,3-benzimidazole

Hazard: Irritant

Formula: C₁₂H₈ClN₃
MW: 229.67 g/mol
MDL Number: MFCD12025828
CAS Number: 143426-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR