2-(2,6-Dimethylmorpholino)-2-methylpropanal ≥95%
Supplier: Matrix Scientific
044014-1G
044014-500MG
044014-5G
101863-680EA
1600
USD
101863-680
101863-676
101863-678
2-(2,6-Dimethylmorpholino)-2-methylpropanal ≥95%
2-(2,6-Dimethylmorpholino)-2-methylpropanal
Hazard: Irritant
Formula:
C₁₀H₁₉NO₂ MW: 185.27 g/mol |
MDL Number:
MFCD08056618 CAS Number: 886361-53-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...