2-Propyl-1,3-benzoxazol-5-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Amino-2-propyl-1,3-benzoxazole

043990-5G 043990-1G 043990-500MG
101863-554EA 556.92 USD
101863-554 101863-558 101863-556
2-Propyl-1,3-benzoxazol-5-amine ≥95%
2-Propyl-1,3-benzoxazol-5-amine

Hazard: Irritant

Formula: C₁₀H₁₂N₂O
MW: 176.22 g/mol
MDL Number: MFCD06200946
CAS Number: 886361-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR