1-[4-(3-Fluoropropoxy)phenyl]-1-ethanone ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(3-Fluoropropoxy)acetophenone

043895-5G 043895-500MG 043895-1G
101863-100EA 231.15 USD
101863-100 101863-098 101863-102
1-[4-(3-Fluoropropoxy)phenyl]-1-ethanone ≥95%
1-[4-(3-Fluoropropoxy)phenyl]-1-ethanone

Hazard: Irritant

Formula: C₁₁H₁₃FO₂
MW: 196.22 g/mol
MDL Number: MFCD03001251
CAS Number: 400878-19-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR