2-Butyl-5-(4-nitrophenyl)-1,3,4-oxadiazole ≥95%

Supplier: Matrix Scientific

043374-1G 043374-5G 043374-500MG
101860-468EA 350.77 USD
101860-468 101860-470 101860-466
2-Butyl-5-(4-nitrophenyl)-1,3,4-oxadiazole ≥95%
2-Butyl-5-(4-nitrophenyl)-1,3,4-oxadiazole

Hazard: Irritant

Formula: C₁₂H₁₃N₃O₃
MDL Number: MFCD06797384
CAS Number: 100933-81-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR