2-Methyl-2-piperidinopropanal ≥95%

Supplier: Matrix Scientific

043370-500MG 043370-5G 043370-1G
101860-456EA 350.77 USD
101860-456 101860-458 101860-460
2-Methyl-2-piperidinopropanal ≥95%
2-Methyl-2-piperidinopropanal

Hazard: Irritant

Formula: C₉H₁₇NO
MW: 155.24 g/mol
MDL Number: MFCD08056609
CAS Number: 16042-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR