4-(4-Pentylcyclohexyl)aniline ≥95%

Supplier: Matrix Scientific

043267-500MG 043267-1G
101859-982EA 349.6 USD
101859-982 101859-980
4-(4-Pentylcyclohexyl)aniline ≥95%
4-(4-Pentylcyclohexyl)aniline

Hazard: Irritant

Formula: C₁₇H₂₇N
MDL Number: MFCD00981976
CAS Number: 73163-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR